missing translation for 'onlineSavingsMsg'
Learn More

Parvovirus VP2 Antibody (4E5A4) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

£197.00 - £435.00

Specifications

Antigen Parvovirus VP2
Clone 4E5A4
Concentration 1 mg/mL
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18255151
View Documents
Novus Biologicals
NBP2-61902
0.1 mL
£435.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18620648
View Documents
Novus Biologicals
NBP2-61902-0.025ml
0.025 mL
£197.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Parvovirus VP2 Monoclonal specifically detects Parvovirus VP2 in Viral samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Parvovirus VP2
1 mg/mL
Monoclonal
Purified
Parvovirus capsid protein 2, Parvovirus major capsid protein
Purified recombinant fragment of human VP2 (AA: 296-438) expressed in E. Coli.
Primary
46.5 kDa
4E5A4
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000
Unconjugated
Mouse
VP2
IgG1
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.