missing translation for 'onlineSavingsMsg'
Learn More

Parvin gamma Mouse anti-Human, Clone: 8C5.2, Novus Biologicals™

Product Code. 18372682 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18372682 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18372682 Supplier Novus Biologicals Supplier No. NBP30783520UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Parvin gamma Monoclonal specifically detects Parvin gamma in Human samples. It is validated for Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Parvin gamma
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 8C5.2
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, Immunoprecipitation 1-2 ug per 100-500 ug of protein 1ml of lysate
Formulation 10 mM PBS with 0.05% BSA
Gene Alias gamma-parvin, parvin, gamma
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human Parvin gamma
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64098
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.