missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PARVG Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583210
This item is not returnable.
View return policy
Description
Immunogen sequence: LHLKEFYLTP NSPAEMLHNV TLALELLKDE GLLSCPVSPE DIVNKDAKST LRVLYGLFCK HTQKAHRDRT PHGAPN Highest antigen sequence indentity to the following orthologs: Rat - 76%, Mouse - 74%.
Probably plays a role in the regulation of cell adhesion and cytoskeleton organization.
Specifications
| PARVG | |
| Polyclonal | |
| Unconjugated | |
| Parvg | |
| AI413459; Gamma-parvin; gamma-parvin; LOW QUALITY PROTEIN: gamma-parvin; PARVG; parvin gamma; parvin, gamma; zgc:162304 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64098 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HBI0 | |
| Parvg | |
| Recombinant protein corresponding to Human PARVG. Recombinant protein control fragment (Product #RP-95368). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction