missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARVB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14463-1-AP-150UL
This item is not returnable.
View return policy
Description
Members of the parvin family, including PARVB, are actin-binding proteins associated with focal contacts.Members of the parvin family, including PARVB, are actin-binding proteins associated with focal contacts.Specifications
| PARVB | |
| Polyclonal | |
| Unconjugated | |
| PARVB | |
| Affixin, Beta parvin, CGI 56, PARVB, parvin, beta | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 170736, 29780, 362973 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9ES46, Q9HBI1 | |
| Parvb | |
| PARVB Fusion Protein Ag5760 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction