missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Parvalbumin Antibody (C12), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79824
This item is not returnable.
View return policy
Description
Parvalbumin Monoclonal specifically detects Parvalbumin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| Parvalbumin | |
| Monoclonal | |
| Unconjugated | |
| PBS pH 7.4, 50% glycerol with 0.09% Sodium Azide | |
| PVALB | |
| Synthetic peptide from the full length Rat Parvalbumin protein | |
| 0.1 mg | |
| Neuroscience | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| C12 | |
| Western Blot, ELISA | |
| D22S749, MGC116759, parvalbumin, parvalbumin alpha, PV | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at -20 °C. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction