missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Parvalbumin alpha Antibody (PVALB/7601) [Allophycocyanin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27001APC
This item is not returnable.
View return policy
Description
Parvalbumin alpha Monoclonal antibody specifically detects Parvalbumin alpha in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Parvalbumin alpha | |
| Monoclonal | |
| APC | |
| D22S749 | |
| Recombinant fragment (around aa1-110) of human Parvalbumin alpha protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| PVALB/7601 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 5816 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction