missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PARVA Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA526326
This item is not returnable.
View return policy
Description
This antibody is predicted to react with rat based on sequence homology.
The protein encoded by this gene is a transcription factor that contains C2H2-type zinc-fingers. It also contains a positive regulatory domain, which has been found in several other zinc-finger transcription factors including those involved in B cell differentiation and tumor suppression. Studies of the mouse counterpart suggest that this protein may be involved in the development of the central nerve system (CNS), as well as in the pathogenesis of neuronal storage disease. Multiple alternatively spliced transcript variants encoding distinct isoforms have been observed.
Specifications
| PARVA | |
| Polyclonal | |
| Unconjugated | |
| PARVA | |
| 2010012A22Rik; 5430400F08Rik; Actopaxin; Actp; AI225929; Alpha-parvin; alpha-parvin-like; AU042898; Calponin-like integrin-linked kinase-binding protein; CH-ILKBP; FLJ10793; FLJ12254; I79_014720; Matrix-remodeling-associated protein 2; MNCb-0301; MXRA2; PARVA; Parvin; parvin alpha; parvin, alpha | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 55742, 57342 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9EPC1, Q9NVD7 | |
| PARVA | |
| KLH conjugated synthetic peptide between 323-351 amino acids from the C-terminal region of human PARVA. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction