missing translation for 'onlineSavingsMsg'
Learn More

3M™ Particulate Filter, P3 R, 2138

Product Code. 15719007
Change view
Click to view available options
:
Particulate Filter 2138, P3 R, 1 pair
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15719007 Particulate Filter 2138, P3 R, 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15719007 Supplier 3M™ Supplier No. 7000029735

Please to purchase this item. Need a web account? Register with us today!

Edge

3M™ Particulate Filters, 2138 connect to any half or full-face 3M™ Reusable Masks with a bayonet-style connection. Our P3 R filters are small sized and lightweight, with low breathing resistance.

For protection against solid and liquid particles, and ozone up to 10 x OEL, get 3M™ Particulate Filters, 2138. They also offer additional protection against organic vapour and acid gases below OEL. Our lightweight, P3 R filters combine protection with comfort. They’re versatile enough to use across our range of reusable half and full-face masks, for safe working in hazardous environments. They offer low breathing resistance and good balance when connected to your mask, and their flexibility means they fit comfortably under welding shields. They can be clicked into place quickly and easily using the 3M™ Bayonet Filter Attachment System, and they can be used with any 3M™ Reusable Masks with a bayonet-style connection.
TRUSTED_SUSTAINABILITY

Specifications

Cartridge Type ‎P3 R
Certifications/Compliance EN143 : 2000, CE Approved
Connection Type Bayonet
Description 10 x TLV and organic vapours/acid gas below TLV, Solid and liquid particles. Plus ozone protection
Color White
Filter Type P3R
Product Type Particulate Filter
For Use With (Application) Pharmaceutical, Powdered chemicals, Construction, Quarrying ceramics, Refractory materials
For Use With (Equipment) Half- and Full-face masks
Quantity 1 Pair
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.