missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ 3M™ Particulate Filter, P3 R, 2135
Description
For breathing protection against solid and liquid particles, use 3M™ Particulate Filters, 2135. They’re versatile enough to use across our range of reusable half and full-face masks, for safe working in hazardous environments. Our lightweight, P3 R filters combine protection with comfort. They offer low breathing resistance and good balance when connected to your mask, and their flexible construction means they fit comfortably under welding shields. They can be clicked into place quickly and easily, and they can be used with any 3M™ Reusable Masks with a bayonet-style connection.
Specifications
Specifications
| Cartridge Type | P3 R |
| Certifications/Compliance | EN143:2000 |
| Connection Type | Bayonet |
| Description | Solid and liquid particles |
| Color | White |
| Filter Type | P3R |
| Product Type | Particulate Filter |
| For Use With (Application) | Pharmaceutical, Powdered chemicals, Construction, Quarrying ceramics, Refractory materials |
| For Use With (Equipment) | Half- and Full-face masks |
| Quantity | 1 Pair |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?