missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Particle-Certified Glass Containers

Product Code. 16484390
Click to view available options
Capacity (Metric):
60 mL
125 mL
250 mL
500 mL
1000 mL
2500 mL
4000 mL
Product Type:
Boston Round Bottle
Jug
Packer Bottle
Sample Jar
Color:
Amber
Clear
No. per Case:
4
12
24
Unit Size:
Each
Pack of 12
Pack of 24
Pack of 4
This item is not returnable. View return policy

Product Code. 16484390

Brand: Thermo Scientific™ 13332C/LP

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Protect the quality of your products or laboratory samples in cleanroom environments by using particle-certified glass containers.

Protect the quality of your products or lab samples. Thermo Scientific™ Particle-Certified Glass Containers are tested and certified to meet the low particle criteria; assembled to contain as few as 5 particles per milliliter > 0.5 microns. A large variant of standard particle-certified containers are available. Non-standard containers may be custom cleaned on request.

Key Features:

  • Class 100/10 glass
  • Certificate of Analysis is included with each shipment
  • Available in clear or amber
  • Assembled with low-shedding polypropylene septa with chemically inert PTFE faced liners that do not contain adhesives
  • Individual bottles double bagged to maintain integrity until point of use
  • 4 L jugs suitable for hazardous shipping in conjunction with combination packaging

Recommended for:

  • Low particle applications
TRUSTED_SUSTAINABILITY

Specifications

Capacity 32 oz., 1000 mL
Capacity (Metric) 1000 mL
Capacity (English) 32 oz.
Color Clear
Description Clear Glass Wide Mouth Jar with Polypropylene Closure, 1000 mL
Diameter (English) 3.75 in.
Material Glass
No. per Case 12
Mouth Wide
Height (Metric) 168.28 mm
Product Type Sample Jar
Height (English) 6.625 in.
Closure Liner PTFE
Closure Material Polypropylene, Low-shedding
Closure Size 89-400 mm
Diameter (Metric) 95.25 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.