missing translation for 'onlineSavingsMsg'
Learn More

Parsaclisib, MedChemExpress

Product Code. p-200101828
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16480597

Brand: Medchem Express HY109068

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Parsaclisib (INCB050465) is a potent, selective and orally active inhibitor of PI3Kδ, with an IC50 of 1 nM at 1 mM ATP. Parsaclisib shows approximately 20000-fold selectivity over other PI3K class I isoforms. Parsaclisib can be used for the research of relapsed or refractory B-cell malignancies.

Parsaclisib (INCB050465) is a potent, selective and orally active inhibitor of PI3Kδ, with an IC50 of 1 nM at 1 mM ATP. Parsaclisib shows approximately 20000-fold selectivity over other PI3K class I isoforms. Parsaclisib can be used for the research of relapsed or refractory B-cell malignancies.

Specifications

Chemical Name or Material Parsaclisib
CAS 1426698-88-5
Color White
Purity Grade Notes Research
Molecular Formula C20H22ClFN6O2
Quantity 50 mg
Synonym INCB050465
Solubility Information DMSO : 125 mg/mL (288.76 mM; Need ultrasonic)
SMILES CCOC1=C([C@@H](C2)CNC2=O)C(F)=C(Cl)C=C1[C@@H](N3C4=NC=NC(N)=C4C(C)=N3)C
Molecular Weight (g/mol) 432.88
Formula Weight 432.88
Percent Purity 98.51%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.