missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PARPYnD

Product Code. 18708346
Change view
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18708346 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18708346 Supplier Tocris Bioscience™ Supplier No. 7410/2
Stock Up and Save This Summer
Use promo code 27950
View all promotions
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

PARP inhibitor; photoaffinity probe

PARPYnD is a potent photoaffinity probe for poly(ADP-ribose) polymerase (PARP) (IC50 values for PARP2, PARP1 and PARP6 are 6, 38 and 230 nM, respectively). PARPYnD labels PARP1 and PARP2 in the cell when an N3 functionalized fluorescent probe is attached and can inhibit isolated PARP6.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(3-Butyn-1-yl)-N-[5-cyano-6-[4-[3-[(3,4-dihydro-4-oxo-1-phthalazinyl)methyl]benzoyl]-1-piperazinyl]-3-pyridinyl]-3 H-diazirine-3-propanamide
CAS 2561483-27-8
Quantity 2 mg
Target Poly(ADP-ribose) Polymerase Inhibitors
Molecular Formula C34H31N9O3
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.