missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARP8 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93551-0.02ml
This item is not returnable.
View return policy
Description
PARP8 Polyclonal antibody specifically detects PARP8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PARP8 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| EC 2.4.2.30, FLJ21308, MGC42864, PARP-8, pART16, poly (ADP-ribose) polymerase family, member 8, poly [ADP-ribose] polymerase 8 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human PARP8 (NP_001171527.1). MGMCSRQERIQKDIDVVIQKSRAEKDCLFADFRYSDSTFTFTYVGGPRSVSYSVHVSEDYPDNTYVSSSENDEDVLVTTEPIPVIFHRIATELRKTNDINCCLSIKSKLQKENGEESRQNSTVEEDSEGDNDSEEFYYGGQVNYDGELHKHPQLEADLSAVREIYGPHAVSLREYGAIDDVDIDLHIDVSFLDEEIAVAWEVIRTEPIIVRLHCSLTQYLNGPVPTVDVFQISTKERFGLGHQLKKIMQTFVTQQWKQSKEKSNCLHNKKLSEKKVKSPLHLFSTLRRSPSYPPPGCGKS | |
| 0.02 mL | |
| Apoptosis | |
| 79668 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction