missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARP4 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97710PCP
This item is not returnable.
View return policy
Description
PARP4 Polyclonal antibody specifically detects PARP4 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| PARP4 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| 193 kDa vault protein, ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase)-like 1, ADPRTL1I-alpha-I-related, EC 2.4.2.30, H5 proline-rich, p193, PARP-4, PARPL, PARP-related, PARP-related/IalphaI-related H5/proline-rich, PH5P, poly (ADP-ribose) polymerase family, member 4, poly [ADP-ribose] polymerase 4, poly(ADP-ribose) synthetase, poly(ADP-ribosyl)transferase-like 1, Vault poly(ADP-ribose) polymerase, vault protein, 193-kDa, VAULT3, von Willebrand factor A domain containing 5C, VPARPKIAA0177, VWA5C | |
| Produced in rabbits immunized with E. coli-derived Human PARP4 fragment. | |
| 0.1 mL | |
| Apoptosis, Cancer, DNA Repair, Hypoxia | |
| 143 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction