missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11289-1-AP-150UL
This item is not returnable.
View return policy
Description
PARP3, poly(ADP-ribosyl)transferase 3, belongs to the PARP family. These enzymes modify nuclear proteins by poly-ADP-ribosylation, which is required for DNA repair, regulation of apoptosis, and maintece of genomic stability. This protein is preferentially localized to the daughter centriole throughout the cell cycle.Specifications
| PARP3 | |
| Polyclonal | |
| Unconjugated | |
| PARP3 | |
| ADPRT 3, ADPRT3, ADPRTL2, ADPRTL3, hPARP 3, IRT1, PADPRT 3, PARP 3, PARP3, Poly [ADP ribose] polymerase 3, Poly[ADP ribose] synthase 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10039, 235587, 300985 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q3ULW8, Q9Y6F1 | |
| PARP3 | |
| PARP3 Fusion Protein Ag1844 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction