missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PARP3 Recombinant Rabbit Monoclonal Antibody (HL1509)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547073
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
PARP3, poly(ADP-ribosyl)transferase 3, belongs to the PARP family. These enzymes modify nuclear proteins by poly-ADP-ribosylation, which is required for DNA repair, regulation of apoptosis, and maintenance of genomic stability. This protein is preferentially localized to the daughter centriole throughout the cell cycle.
Specifications
| PARP3 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9Y6F1 | |
| PARP3 | |
| Recombinant protein encompassing a sequence within the center region of human PARP3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL1509 | |
| Unconjugated | |
| PARP3 | |
| A DP-ribosyltransferase (NAD+; poly (ADP-ribose polymerase)-like 3; A930002C11Rik; ADP-ribosyltransferase (NAD+, poly (ADP-ribose polymerase)-like 3; ADP-ribosyltransferase (NAD+; poly (ADP-ribose polymerase)-like 3; ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase)-like 2; ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase)-like 3; ADP-ribosyltransferase diphtheria toxin-like 3; Adprt3; ADPRT-3; ADPRTL2; Adprtl3; ARTD3; AW990611; DNA ADP-ribosyltransferase PARP3; IRT1; NAD(+) ADP-ribosyltransferase 3; NAD+ ADP-ribosyltransferase 3; pADPRT-3; Parp3; PARP-3; poly (ADP-ribose) polymerase family, member 3; poly (ADP-ribosyl) transferase-like 3; poly [ADP-ribose] polymerase 3; poly(ADP-ribose) polymerase family member 3; poly(ADP-ribose) synthetase-3; poly[ADP-ribose] synthase 3; poly[ADP-ribose] synthetase 3; Poly[ADP-ribose] synthetase-3; Protein mono-ADP-ribosyltransferase PARP3 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10039, 235587 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction