missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARP3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £375.00
Specifications
| Antigen | PARP3 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50-1:100 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18696961
|
Novus Biologicals
NBP2-93033-0.02ml |
0.02 mL |
£161.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18626590
|
Novus Biologicals
NBP2-93033-0.1ml |
0.1 mL |
£375.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PARP3 Polyclonal antibody specifically detects PARP3 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| PARP3 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| PBS (pH 7.3), 50% glycerol | |
| 10039 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50-1:100 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase)-like 3, ADPRT-3, ADPRT3PADPRT-3, ADPRTL2, ADPRTL3NAD+ ADP-ribosyltransferase 3, EC 2.4.2.30, hPARP-3, IRT1ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase)-like 2, NAD(+) ADP-ribosyltransferase 3, pADPRT-3, PARP-3, poly (ADP-ribose) polymerase family, member 3, poly [ADP-ribose] polymerase 3, poly(ADP-ribose) synthetase-3, Poly[ADP-ribose] synthase 3, poly[ADP-ribose] synthetase 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 301-540 of human PARP3 (NP_001003931.3). LAQALQAVSEQEKTVEEVPHPLDRDYQLLKCQLQLLDSGAPEYKVIQTYLEQTGSNHRCPTLQHIWKVNQEGEEDRFQAHSKLGNRKLLWHGTNMAVVAAILTSGLRIMPHSGGRVGKGIYFASENSKSAGYVIGMKCGAHHVGYMFLGEVALGREHHINTDNPSLKSPPPGFDSVIARGHTEPDPTQDTELELDGQQVVVPQGQPVPCPEFSSSTFSQSEYLIYQESQCRLRYLLEVHL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title