missing translation for 'onlineSavingsMsg'
Få mere at vide

PARP3 Antibody (2H6), Novus Biologicals™

Artikelnummer. 18357569 Shop alle Bio Techne produkter
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
0.1 mg
Pakningsstørrelse:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
18357569 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18357569 Leverandør Novus Biologicals Leverandørnr. H00010039M06

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Mouse Monoclonal Antibody

PARP3 Monoclonal antibody specifically detects PARP3 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen PARP3
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2H6
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH14260
Gene Alias ADP-ribosyltransferase (NAD+, poly (ADP-ribose) polymerase)-like 3, ADPRT-3, ADPRT3PADPRT-3, ADPRTL2, ADPRTL3NAD+ ADP-ribosyltransferase 3, EC 2.4.2.30, hPARP-3, IRT1ADP-ribosyltransferase (NAD+, poly (ADP-ribose) polymerase)-like 2, NAD(+) ADP-ribosyltransferase 3, pADPRT-3, PARP-3, poly (ADP-ribose) polymerase family, member 3, poly [ADP-ribose] polymerase 3, poly(ADP-ribose) synthetase-3, Poly[ADP-ribose] synthase 3, poly[ADP-ribose] synthetase 3
Host Species Mouse
Immunogen PARP3 (AAH14260, 1 a.a. ~ 533 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAPKPKPWVQTEGPEKKKGRQAGREEDPFRSTAEALKAIPAEKRIIRVDPTCPLSSNPGTQVYEDYNCTLNQTNIENNNNKFYIIQLLQDSNRFFTCWNRWGRVGEVGQSKINHFTRLEDAKKDFEKKFREKTKNNWAERDHFVSHPGKYTLIEVQAEDEAQEAVVKVDRGPVRTVTKRVQPCSLDPATQKLITNIFSKEMFKNTMALMDLDVKKMPLGKLSKQQIARGFEALEALEEALKGPTDGGQSLEELSSHFYTVIPHNFGHSQPPPINSPELLQAKKDMLLVLADIELAQALQAVSEQEKTVEEVPHPLDRDYQLLKCQLQLLDSGAPEYKVIQTYLEQTGSNHRCPTLQHIWKVNQEGEEDRFQAHSKLGNRKLLWHGTNMAVVAAILTSGLRIMPHSGGRVGKGIYFASENSKSAGYVIGMKCGAHHVGYMFLGEVALGREHHINTDNPSLKSPPPGFDSVIARGHTEPDPTQDTELELDGQQVVVPQGQPVPCPEFSSSTFSQSEYLIYQESQCRLRYLLEVHL
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 10039
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.