missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARP16 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06231PE
This item is not returnable.
View return policy
Description
PARP16 Polyclonal antibody specifically detects PARP16 in Human samples. It is validated for Western Blot
Specifications
| PARP16 | |
| Polyclonal | |
| Western Blot | |
| C15orf30, chromosome 15 open reading frame 30, EC 2.4.2.30, FLJ20509, FLJ25281, PARP-16, pART15, poly (ADP-ribose) polymerase family, member 16, poly [ADP-ribose] polymerase 16 | |
| Produced in rabbits immunized with E. coli-derived Human PARP16 fragment. | |
| 0.1 mL | |
| Apoptosis | |
| 54956 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| PE | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction