missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PARP16 Polyclonal specifically detects PARP16 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | PARP16 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 650 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | C15orf30, chromosome 15 open reading frame 30, EC 2.4.2.30, FLJ20509, FLJ25281, PARP-16, pART15, poly (ADP-ribose) polymerase family, member 16, poly [ADP-ribose] polymerase 16 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human PARP16 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?