missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARP11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16692-1-AP-150UL
This item is not returnable.
View return policy
Description
Poly(ADP-ribosylation) is a method of DNA damage-dependent posttranslational modification that helps to rescue injured proliferating cells from cell death. The PARP (poly(ADP-ribose) polymerase) proteins comprise a superfamily of enzymes that functionally modify histones and other nuclear proteins, thereby preventing cell death. PARPs use NAD+ as a substrate to catalytically transfer ADP-ribose residues onto protein acceptors; a process that, when repeated multiple times, leads to the formation of poly(ADPribose) chains on the protein.Specifications
| PARP11 | |
| Polyclonal | |
| Unconjugated | |
| PARP11 | |
| C12orf6, PARP 11, PARP11 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 101187, 500323, 57097 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8CFF0, Q9NR21 | |
| Parp11 | |
| PARP11 Fusion Protein Ag10053 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction