missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PARP11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550474
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total PARP11 protein.
Mono-ADP-ribosyltransferase that mediates mono-ADP-ribosylation of target proteins (PubMed:25043379, PubMed:25673562). Plays a role in nuclear envelope stability and nuclear remodeling during spermiogenesis. [UniProt].
Specifications
| PARP11 | |
| Polyclonal | |
| Unconjugated | |
| Parp11 | |
| 5330431N24Rik; ADP-ribosyltransferase diphtheria toxin-like 11; AI851877; ARTD11; C12orf6; HIN1L; MIB006; PARP11; PARP-11; poly (ADP-ribose) polymerase family, member 11; poly [ADP-ribose] polymerase 11; poly(ADP-ribose) polymerase family member 11; Protein mono-ADP-ribosyltransferase PARP11; RGD1561791 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57097 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.7 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9NR21 | |
| Parp11 | |
| Full length fusion protein of Human PARP11. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction