Learn More
PARP10 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26072-1-AP-150UL
 View more versions of this product
Product Code. 16874595
  • £349.00 / 150µL
Description
E3 ubiquitin-protein ligase which is a component of the N-end rule pathway. Recognizes and binds to proteins bearing specific amino-terminal residues that are destabilizing according to the N-end rule, leading to their ubiquitination and subsequent degradation.
Specifications
Specifications
PARP10
Polyclonal
Unconjugated
PARP10
PARP 10, PARP10
Rabbit
Antigen Affinity Chromatography
RUO
84875
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.39 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q53GL7
PARP10
PARP10 Fusion Protein Ag23457
150 μL