missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PARP10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542250
This item is not returnable.
View return policy
Description
Immunogen sequence: PDDPSGLPGR SPDT Sequence homology: Human: 100%.
Poly(ADP-ribose) polymerases (PARPs), such as PARP10, regulate gene transcription by altering chromatin organization by adding ADP-ribose to histones. PARPs can also function as transcriptional cofactors. Poly(ADP-ribose) polymerases (PARPs), such as PARP10, regulate gene transcription by altering chromatin organization by adding ADP-ribose to histones. PARPs can also function as transcriptional cofactors (Yu et al., 2005).
Specifications
| PARP10 | |
| Polyclonal | |
| Unconjugated | |
| PARP10 | |
| ADP-ribosyltransferase diphtheria toxin-like 10; ARTD10; FLJ14464; LOC102548778; Parp10; PARP-10; poly (ADP-ribose) polymerase family, member 10; poly [ADP-ribose] polymerase 10; poly [ADP-ribose] polymerase 10-like; poly(ADP-ribose) polymerase family member 10; Protein mono-ADP-ribosyltransferase PARP10 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 84875 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q53GL7 | |
| PARP10 | |
| Synthetic peptide directed towards the C-terminal region of human PARP10 (aa 1012-1025). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction