missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARP1, Rabbit anti-Human, Clone: H8, Abnova™
Rabbit Monoclonal Antibody
Brand: Abnova MAB18755.20uL
This item is not returnable.
View return policy
Specifications
| PARP1 | |
| Monoclonal | |
| Unconjugated | |
| In PBS, pH 7.4 (50% glycerol, 0.1% BSA and 0.02% sodium azide). | |
| ADPRT/ADPRT1/PARP/PARP-1/PPOL/pADPRT-1 | |
| Rabbit | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry, Western Blot | |
| H8 | |
| Flow Cytometry Western Blot The optimal working dilution should be determined by the end user. | |
| poly (ADP-ribose) polymerase 1 | |
| PARP1 | |
| A synthetic peptide corresponding to residues surrounding D214 of human PARP1. | |
| RUO | |
| 142 | |
| Store at -20°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction