missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARP Rabbit anti-Human, Mouse, Clone: 4M9X4, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £395.00
Specifications
| Antigen | PARP |
|---|---|
| Clone | 4M9X4 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18388544
|
Novus Biologicals
NBP3-15813-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18346705
|
Novus Biologicals
NBP3-15813-100UL |
100 μg |
£395.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PARP Monoclonal antibody specifically detects PARP in Human, Mouse samples. It is validated for Western BlotSpecifications
| PARP | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Base Excision Repair, Cancer, Core ESC Like Genes, DNA Repair, Hypoxia, Necroptosis, Stem Cell Markers, Tumor Suppressors | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 142 | |
| IgG | |
| Affinity purified |
| 4M9X4 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase), ADPRT 1, ADPRTADP-ribosyltransferase NAD(+), EC 2.4.2, EC 2.4.2.30, NAD(+) ADP-ribosyltransferase 1, PARP apoptosis, PARP-1, PARPADPRT1, poly (ADP-ribose) polymerase 1, poly (ADP-ribose) polymerase family, member 1, poly [ADP-ribose] polymerase 1, poly(ADP-ribose) polymerase, poly(ADP-ribose) synthetase, poly(ADP-ribosyl)transferase, Poly[ADP-ribose] synthase 1, PPOLpADPRT-1 | |
| A synthetic peptide corresponding to a sequence within amino acids 150-250 of human PARP (P09874). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title