missing translation for 'onlineSavingsMsg'
Learn More

Paroxypropione, MedChemExpress

Product Code. 16492329
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492329 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16492329 Supplier Medchem Express Supplier No. HYB1353

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Paroxypropione is a manufactured, nonsteroidal estrogen which has been used medically as an antigonadotropin.

Paroxypropione is a manufactured, nonsteroidal estrogen which has been used medically as an antigonadotropin.

Specifications

Chemical Name or Material Paroxypropione
CAS 70-70-2
Color White
Purity Grade Notes Research
Molecular Formula C9H10O2
Quantity 1 g
Synonym 4'-Hydroxypropiophenone
Solubility Information DMSO : 100 mg/mL (665.91 mM; Need ultrasonic and warming)
SMILES CCC(C1=CC=C(O)C=C1)=O
Molecular Weight (g/mol) 150.17
Formula Weight 150.17
Percent Purity 99.54%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.