missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Paroxetine hydrochloride, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
Paroxetine hydrochloride is a potent selective serotonin-reuptake inhibitor, commonly prescribed as an and has GRK2 inhibitory ability with IC50 of 14 μM. Paroxetine hydrochloride can be used for the research of depressive disorder.
Specifications
Specifications
| Chemical Name or Material | Paroxetine hydrochloride |
| CAS | 78246-49-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C19H21ClFNO3 |
| Quantity | 100 mg |
| Synonym | BRL29060 hydrochloride BRL29060A |
| Solubility Information | DMSO : 100 mg/mL (273.35 mM; Need ultrasonic) ∣H2O : 5 mg/mL (13.67 mM; Need ultrasonic) |
| SMILES | FC1=CC=C([C@H]2[C@H](COC3=CC=C(OCO4)C4=C3)CNCC2)C=C1.Cl |
| Molecular Weight (g/mol) | 365.83 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction