missing translation for 'onlineSavingsMsg'
Learn More

Paroxetine hydrochloride, MedChemExpress

Product Code. 16472156
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16472156

Brand: Medchem Express HYB0492

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Paroxetine hydrochloride is a potent selective serotonin-reuptake inhibitor, commonly prescribed as an and has GRK2 inhibitory ability with IC50 of 14 μM. Paroxetine hydrochloride can be used for the research of depressive disorder.

Paroxetine hydrochloride is a potent selective serotonin-reuptake inhibitor, commonly prescribed as an and has GRK2 inhibitory ability with IC50 of 14 μM. Paroxetine hydrochloride can be used for the research of depressive disorder.

Specifications

Chemical Name or Material Paroxetine hydrochloride
CAS 78246-49-8
Color White
Purity Grade Notes Research
Molecular Formula C19H21ClFNO3
Quantity 500 mg
Synonym BRL29060 hydrochloride BRL29060A
Solubility Information DMSO : 100 mg/mL (273.35 mM; Need ultrasonic) ∣H2O : 5 mg/mL (13.67 mM; Need ultrasonic)
SMILES FC1=CC=C([C@H]2[C@H](COC3=CC=C(OCO4)C4=C3)CNCC2)C=C1.Cl
Molecular Weight (g/mol) 365.83
Formula Weight 365.83
Percent Purity 99.92%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.