missing translation for 'onlineSavingsMsg'
Learn More

Paroxetine Hydrochloride Hemihydrate, LoGiCal

Product Code. 30461369
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30461369 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461369 Supplier LoGiCal Supplier No. LGCFOR0264.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Spécification

Chemical Name or Material Paroxetine Hydrochloride Hemihydrate
CAS 110429-35-1
Analyte or Component Names Paroxetine Hydrochloride Hemihydrate
Shipping Condition Room Temperature
Molecular Formula 2 C19 H20 F N O3 . 2 Cl H . H2 O
Certifications/Compliance ISO 9001
Synonym Paroxetine Hydrochloride Hemihydrate, Piperidine, 3-[(1,3-benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)-, hydrochloride, hydrate (2:2:1), (3S,4R)-, Piperidine, 3-[(1,3-benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)-, hydrochloride, hydrate (2:1), (3S,4R)- (9CI), Piperidine, 3-[(1,3-benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)-, hydrochloride, hydrate (2:1), (3S-trans)-, (-)-trans-4-(4-Fluorophenyl)-3-(3,4-methylenedioxyphenoxymethyl)piperidine hydrochloride hemihydrate, Paroxetine hydrochloride hemihydrate, BRL-29060A, Form 1 Pseudopolymorph, Paroxetine hydrochloride hemihydrate (BRL29060A), BP Paroxetine Impurity Standard, Spiked with Ph Eur Paroxetine Hydrochloride Impurity G, BRL-55490 (F-THP)
InChI Formula InChI=1S/2C19H20FNO3.2ClH.H2O/c2*20-15-3-1-13(2-4-15)17-7-8-21-10-14(17)11-22-16-5-6-18-19(9-16)24-12-23-18;;;/h2*1-6,9,14,17,21H,7-8,10-12H2;2*1H;1H2/t2*14-,17-;;;/m00.../s1
Solution Type Neat
SMILES O.Cl.Cl.Fc1ccc(cc1)[C@@H]2CCNC[C@H]2COc3ccc4OCOc4c3.Fc5ccc(cc5)[C@@H]6CCNC[C@H]6COc7ccc8OCOc8c7
IUPAC Name (3S,4R)-3-(1,3-benzodioxol-5-yloxymethyl)-4-(4-fluorophenyl)piperidine;hydrate;dihydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 749.67
Quantity 10 mg
Formula Weight 748.2494 g/mol
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.