missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Paroxetine EP Impurity B, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
Paroxetine EP Impurity B
Specifications
Specifications
| Chemical Name or Material | Sesamol |
| CAS | 533-31-3 |
| Analyte or Component Names | Sesamol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C7 H6 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 1,3-Benzodioxol-5-ol (Sesamol), Paroxetine Hydrochloride Anhydrous Imp. B (EP), Paroxetine Imp. B (EP), 1,3-Benzodioxol-5-ol, Sesamol, BRL-43782, Paroxetine Related, 3,4-Methylenedioxyphenol, Sesamol (BRL43782), BP Paroxetine Impurity Standard, Paroxetine Hydrochloride Impurity B, Paroxetine Impurity B |
| InChI Formula | InChI=1S/C7H6O3/c8-5-1-2-6-7(3-5)10-4-9-6/h1-3,8H,4H2 |
| SMILES | Oc1ccc2OCOc2c1 |
| IUPAC Name | 1,3-benzodioxol-5-ol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction