missing translation for 'onlineSavingsMsg'
Learn More

Paromomycin Sulfate, Fisher BioReagents

Product Code. 10094487
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10094487 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10094487 Supplier Fisher BioReagents Supplier No. BP29531

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibits protein synthesis

  • A broad spectrum aminoglycoside antibiotic
  • Functions as a protein synthesis inhibitor
  • Useful in the study of bacterial protein synthesis, RNA molecules, and the effects of antibiotics on cell lines
  • Suitable for cell culture applications; research only
TRUSTED_SUSTAINABILITY

Specifications

Type Paromomycin Sulfate
Physical Form Powder/Solid
CAS 1263-89-4
Source Streptomyces krestomuceticus
Loss on Drying < 5%
Packaging Amber Glass
Potency > 675 U/mg
Quantity 1 g
Molecular Formula C23H47N5O18S
InChI Key LJRDOKAZOAKLDU-JXDXLELXSA-N
SMILES C1C(C(C(C(C1N)OC2C(C(C(C(O2)CO)O)O)N)OC3C(C(C(O3)CO)OC4C(C(C(C(O4)CN)O)O)N)O)O)N.OS(=O)(=O)O
IUPAC Name (3S,5S,6S)-5-amino-2-(aminomethyl)-6-[(2R,4S,5S)-5-[(2R,5R,6R)-3,5-diamino-2-[(2S,3S,5S)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-hydroxycyclohexyl]oxy-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]oxyoxane-3,4-diol;sulfuric acid
Molecular Weight (g/mol) 713.706
PubChem CID 91972180
Formula Weight 615.6
Chemical Name or Material Paromomycin Sulfate
Show More Show Less
compliance-icons
Product Identifier
  • Paromomycin sulfate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.