missing translation for 'onlineSavingsMsg'
Learn More

Paromomycin sulfate, 98.3%, MP Biomedicals™

Product Code. 11489381 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11489381 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11489381 Supplier MP Biomedicals Supplier No. 0215605605
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Broad-spectrum aminoglycoside antibiotic used to study bacterial protein synthesis and 30S ribosome assembly.

Specifications

Type Paromomycin Sulfate
Physical Form Powder
CAS 149-29-1, 7732-18-5
Color Off-White
Quantity 5 g
Assay Percent Range >97%
Molecular Formula C7H6O4
InChI Key ZRWPUFFVAOMMNM-UHFFFAOYNA-N
SMILES OC1OCC=C2OC(=O)C=C12
IUPAC Name 4-hydroxy-2H,4H,6H-furo[3,2-c]pyran-2-one
Molecular Weight (g/mol) 154.12
Formula Weight 615.6
Percent Purity >97%
Grade Not Available
Chemical Name or Material Paromomycin Sulfate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.