missing translation for 'onlineSavingsMsg'
Learn More

Paromomycin Deuterated Acetic Acid Salt, TRC

Product Code. 30425439
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30425439 10 mg 10mg
30417869 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30425439 Supplier Toronto Research Chemicals Supplier No. TRCP20565310MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Paromomycin Deuterated Acetic Acid Salt
Molecular Formula C23H45N5O14•5(CH3COOH) for undeuterated
Quantity 10 mg
Synonym O-2,6-Diamino-2,6-dideoxy-β-L-idopyranosyl-(1→3)-O-β-D-ribofuranosyl-(1→5)-O-[2-amino-2-deoxy-α-D-glucopyranosyl-(1→4)]-2-deoxy-D-Streptamine Deuterated Acetic Acid Salt, O-2-Amino-2-deoxy-α-D-glucopyranosyl-(1→4)-O-[O-2,6-diamino-2,6-dideoxy-β-L-idopyranosyl-(1→3)-β-D-ribofuranosyl-(1→5)]-2-deoxy-D-streptamine Deuterated Acetic Acid Salt, 1600 Antibiotic Deuterated Acetic Acid Salt, Aminosidin Deuterated Acetic Acid Salt, Aminosidine Deuterated Acetic Acid Salt, Aminoxidin Deuterated Acetic Acid Salt, FI 5853 Deuterated Acetic Acid Salt, Farmiglucin Deuterated Acetic Acid Salt, Farminosidin Deuterated Acetic Acid Salt, Gabbroral Deuterated Acetic Acid Salt, Humagel Deuterated Acetic Acid Salt, Humatin Deuterated Acetic Acid Salt, Humycin Deuterated Acetic Acid Salt, Paramicina Deuterated Acetic Acid Salt, Pargonyl Deuterated Acetic Acid Salt, Paricina Deuterated Acetic Acid Salt, Sinosid Deuterated Acetic Acid Salt
InChI Formula InChI=1S/C23H45N5O14/c24-2-7-13(32)15(34)10(27)21(37-7)41-19-9(4-30)39-23(17(19)36)42-20-12(31)5(25)1-6(26)18(20)40-22-11(28)16(35)14(33)8(3-29)38-22/h5-23,29-36H,1-4,24-28H2/t5-,6+,7+,8-,9-,10-,11-,12+,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23+/m1/s1
SMILES O[C@@H]1[C@H](O[C@H]2O[C@@H](CN)[C@@H](O)[C@H](O)[C@H]2N)[C@@H](CO)O[C@H]1O[C@H]([C@@H](O)[C@H](N)C[C@@H]3N)[C@@H]3O[C@@H]4[C@H](N)[C@@H](O)[C@H](O)[C@@H](CO)O4
Molecular Weight (g/mol) 615.63
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.