missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PARN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115722
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PARN.
The protein encoded by this gene is a 3'-exoribonuclease, with similarity to the RNase D family of 3'-exonucleases. It prefers poly(A) as the substrate, hence, efficiently degrades poly(A) tails of mRNAs. Exonucleolytic degradation of the poly(A) tail is often the first step in the decay of eukaryotic mRNAs. This protein is also involved in silencing of certain maternal mRNAs during oocyte maturation and early embryonic development, as well as in nonsense-mediated decay (NMD) of mRNAs that contain premature stop codons. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| PARN | |
| Polyclonal | |
| Unconjugated | |
| Parn | |
| 1200003I18Rik; DAN; Deadenylating nuclease; deadenylation nuclease; DKCB6; Parn; PFBMFT4; poly(A)-specific ribonuclease; poly(A)-specific ribonuclease (deadenylation nuclease); poly(A)-specific ribonuclease PARN; Polyadenylate-specific ribonuclease; RGD1565449 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 360464, 5073, 74108 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| O95453, Q8VDG3 | |
| Parn | |
| A synthesized peptide derived from human PARN(Accession O95453), corresponding to amino acid residues F8-K58. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction