missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARN Antibody (OTI4D12), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73246PCP
This item is not returnable.
View return policy
Description
PARN Monoclonal antibody specifically detects PARN in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| PARN | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 5073 | |
| Store at 4C in the dark. | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| OTI4D12 | |
| Western Blot, Immunohistochemistry | |
| DANEC 3.1.13.4, Deadenylating nuclease, Deadenylation nuclease, poly(A)-specific ribonuclease (deadenylation nuclease), poly(A)-specific ribonuclease PARN, Polyadenylate-specific ribonuclease | |
| Human recombinant protein fragment corresponding to amino acids 1-266 of human PARN (NP_002573) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction