missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PARL Monoclonal Antibody (4F4)
GREENER_CHOICE

Product Code. 15519082
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15519082 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15519082 Supplier Invitrogen™ Supplier No. MA522148

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Peptide Sequence: AWRGWAQRGW GCGQAWGASV GGRSCEELTA VLTPPQLLGR RFNFFIQQKC GFRKAPRKVE PRRSDPGTSG EAYKRSALIP PVEETVFYPS PYPIRSLIK.

This gene encodes a mitochondrial integral membrane protein. Following proteolytic processing of this protein, a small peptide is formed and translocated to the nucleus. This gene may be involved in signal transduction via regulated intramembrane proteolysis of membrane-tethered precursor proteins. Variation in this gene has been associated with increased risk for type 2 diabetes. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PARL
Applications ELISA
Classification Monoclonal
Clone 4F4
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene PARL
Gene Accession No. Q9H300
Gene Alias D16Ertd607e; Mitochondrial intramembrane cleaving protease PARL; mitochondrial intramembrane-cleaving protease PARL; Parl; parl {ECO:0000250; Pbeta; P-beta; presenilin associated rhomboid like; presenilin associated, rhomboid-like; presenilin associated, rhomboid-like isoform 1 preproprotein; presenilin associated, rhomboid-like preproprotein; presenilins-associated rhomboid-like protein, mitochondrial; PRO2207; Psarl; PSARL1; PSENIP2; RHBDS1; rhomboid 7 homolog 1; UniProtKB:Q9H300}
Gene Symbols PARL
Host Species Mouse
Immunogen PARL (NP_061092.3, 2 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55486
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.