missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARL Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76938AF750
This item is not returnable.
View return policy
Description
PARL Polyclonal antibody specifically detects PARL in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| PARL | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 750 | |
| EC 3.4.21.105, mitochondrial, presenilin associated, rhomboid-like, PRO2207, rhomboid 7 homolog 1 | |
| Antibody was raised against a 15 amino acid synthetic peptide from near the amino terminus of human PARL. The immunogen is located within amino acids 40 - 90 of PARL. Amino Acid Squence: RKAPRKVEPRRSDPG | |
| 0.1 mL | |
| Alzheimers Research, Apoptosis, Mitochondrial Fusion Proteins, Neuroscience | |
| 55486 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction