Learn More
Invitrogen™ Parkin Chimeric Recombinant Mouse Monoclonal Antibody (21H24L9), Invitrogen™
Mouse Recombinant Monoclonal Antibody
Brand: Invitrogen™ 702785M
Description
This antibody has Rabbit Variable heavy and light chains, and Mouse constant domains. It recognizes human, mouse and Rat Parkin protein. This chimeric antibody is recognized by anti-mouse secondary antibodies and has no cross reactivity to anti-rabbit secondary antibodies. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
The precise function of Parkin gene is unknown; however, the encoded protein is a component of a multiprotein E3 ubiquitin ligase complex that mediates the targeting of substrate proteins for proteasomal degradation. Mutations in this gene are known to cause Parkinson disease and autosomal recessive juvenile Parkinson disease. Alternative splicing of this gene produces multiple transcript variants encoding distinct isoforms. Additional splice variants of this gene have been described but currently lack transcript support.
Specifications
| Parkin Chimeric | |
| Recombinant Monoclonal | |
| 1.0 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O60260, Q9JK66, Q9WVS6 | |
| PRKN | |
| Protein corresponding to human Parkin [aa1-aa465]. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| 21H24L9 | |
| Unconjugated | |
| PRKN | |
| AAN12155.1; AR-JP; CG10523; CG10523-PB; CG10523-PC; Dmel\CG10523; Dmel_CG10523; Dpark; dParkin; D-parkin; dpk; E3 ubiquitin ligase; E3 ubiquitin-protein ligase parkin; LPRS2; OTTHUMP00000017562; OTTHUMP00000017563; OTTHUMP00000017564; park; park1; PARK2; Parkin; parkin 2; parkin F; Parkin isoform 1; parkin protein; parkin RBR E3 ubiquitin protein ligase; Parkin RBR E3 ubiquitin-protein ligase; parkin variant SV5DEL; Parkinson disease (autosomal recessive, juvenile) 2, parkin; Parkinson disease protein 2; Parkinson juvenile disease protein 2; parkinson protein 2 E3 ubiquitin protein ligase; parkinson protein 2, E3 ubiquitin protein ligase; parkinson protein 2, E3 ubiquitin protein ligase (parkin); park-PB; park-PC; PDJ; PK; Prkn; SD01679 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 5071, 50873, 56816 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.