Learn More
Parker domnick hunter™ Nylon Deionizer Bags for Hydrogen Gas Generators
Product Code. 12842636
Click to view available options
Type:
Deionizer Bag
Unit Size:
Pack of 2
Product Code. 12842636
Brand: Parker domnick hunter™ 604970010
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Bead color change from gold to green-gray indicates the need for a new bag
  • Contain impregnated silica gel beads to deionize water poured into the generator
  • Bead color change from gold to green-gray indicates the need for a new bag
  • Bags must be used at all times
  • Average bag life of six months can be extended by using high-quality water
  • Fits domnick hunter™ UHP Hydrogen generator models
Order Info
Sold in packs of two.
Specifications
For Use With (Equipment) Domnick hunter uhp hydrogen generator model
Material Nylon
Type Deionizer Bag