missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARK7/DJ-1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11681-1-AP-150UL
This item is not returnable.
View return policy
Description
The product of this gene belongs to the peptidase C56 family of proteins. It acts as a positive regulator of androgen receptor-dependent transcription. It may also function as a redox-sensitive chaperone, as a sensor for oxidative stress, and it apparently protects neurons against oxidative stress and cell death. Defects in this gene are the cause of autosomal recessive early-onset Parkinson disease 7. Two transcript variants encoding the same protein have been identified for this gene.Specifications
| PARK7/DJ-1 | |
| Polyclonal | |
| Unconjugated | |
| PARK7 | |
| DJ 1, DJ1, Oncogene DJ1, PARK7, Parkinson disease protein 7, Protein DJ 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11315, 57320 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.55 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q99497, Q99LX0 | |
| Park7 | |
| PARK7,DJ-1 Fusion Protein Ag2287 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction