missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Park7/DJ-1 Mouse anti-Human, Clone: 1.00E+12, Novus Biologicals™
Mouse Monoclonal Antibody
£241.00 - £378.00
Specifications
| Antigen | Park7/DJ-1 |
|---|---|
| Clone | 1000000000000 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Flow Cytometry 1:10-1:1017, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18203492
|
Novus Biologicals
NBP2-59407-50UL |
50 μL |
£241.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18230404
|
Novus Biologicals
NBP2-59407-100UL |
100 μL |
£378.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Park7/DJ-1 Monoclonal specifically detects Park7/DJ-1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Park7/DJ-1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Alzheimers Research, Apoptosis, Cancer, DNA replication Transcription Translation and Splicing, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Tumor Suppressors | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| 11315 | |
| Recombinant human Park7/DJ-1 (1-189aa) purified from E. coli | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1000000000000 | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1017, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| DJ-1, DJ1FLJ34360, EC 3.4, FLJ27376, FLJ92274, Oncogene DJ1, Parkinson disease (autosomal recessive, early onset) 7, Parkinson disease protein 7, protein DJ-1 | |
| PARK7 | |
| IgG2b κ | |
| Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title