missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Park7/DJ-1 Antibody (4H4), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-92715G
This item is not returnable.
View return policy
Description
Park7/DJ-1 Monoclonal specifically detects Park7/DJ-1 in Human, Bovine, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated.
Specifications
| Park7/DJ-1 | |
| Monoclonal | |
| DyLight 488 | |
| Q99497 | |
| PARK7 | |
| Full length recombinant human Park7(DJ-1) expressed in and purified from E. coli. [UniProt# Q99497] | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Bovine and Mouse. | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoassay | |
| 4H4 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated | |
| DJ-1, DJ1FLJ34360, EC 3.4, FLJ27376, FLJ92274, Oncogene DJ1, Parkinson disease (autosomal recessive, early onset) 7, Parkinson disease protein 7, protein DJ-1 | |
| Mouse | |
| 21 kDa | |
| 0.1 mL | |
| Alzheimers Research, Apoptosis, Cancer, DNA replication Transcription Translation and Splicing, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Tumor Suppressors | |
| 11315 | |
| Human, Bovine, Mouse (Negative), Rat (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction