missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Park7/DJ-1 Monoclonal antibody specifically detects Park7/DJ-1 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | Park7/DJ-1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 1000000000000 |
| Conjugate | mFluor Violet 500 SE |
| Dilution | Flow Cytometry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | DJ-1, DJ1FLJ34360, EC 3.4, FLJ27376, FLJ92274, Oncogene DJ1, Parkinson disease (autosomal recessive, early onset) 7, Parkinson disease protein 7, protein DJ-1 |
| Host Species | Mouse |
| Immunogen | Recombinant human Park7/DJ-1 (1-189aa) purified from E. coli |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?