missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Park7/DJ-1 Antibody (007), Novus Biologicals™
Rabbit Monoclonal Antibody
£214.00 - £353.00
Specifications
| Antigen | Park7/DJ-1 |
|---|---|
| Clone | 007 |
| Dilution | Western Blot 1:1000-1:5000, Immunocytochemistry/ Immunofluorescence 1:20-1:100, Immunoprecipitation 1-4 uL/mg of lysate |
| Applications | Western Blot, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18645478
|
Novus Biologicals
NBP3-05805-100ul |
100 μg |
£353.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18665808
|
Novus Biologicals
NBP3-05805-50ul |
50 μg |
£214.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Park7/DJ-1 Monoclonal antibody specifically detects Park7/DJ-1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
| Park7/DJ-1 | |
| Western Blot 1:1000-1:5000, Immunocytochemistry/ Immunofluorescence 1:20-1:100, Immunoprecipitation 1-4 uL/mg of lysate | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| DJ-1, DJ1FLJ34360, EC 3.4, FLJ27376, FLJ92274, Oncogene DJ1, Parkinson disease (autosomal recessive, early onset) 7, Parkinson disease protein 7, protein DJ-1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Park7/DJ-1 (Uniprot#: Q99497-1; Met1-Asp189). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 007 | |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Alzheimers Research, Apoptosis, Cancer, DNA replication Transcription Translation and Splicing, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Tumor Suppressors | |
| 0.2 um filtered solution in PBS | |
| 11315 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title