missing translation for 'onlineSavingsMsg'
Learn More

Park7/DJ-1 Antibody (007), DyLight 680, Novus Biologicals™

Product Code. 18407819 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407819 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407819 Supplier Novus Biologicals Supplier No. NBP305805FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Park7/DJ-1 Monoclonal antibody specifically detects Park7/DJ-1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Park7/DJ-1
Applications Western Blot, Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 007
Conjugate DyLight 680
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias DJ-1, DJ1FLJ34360, EC 3.4, FLJ27376, FLJ92274, Oncogene DJ1, Parkinson disease (autosomal recessive, early onset) 7, Parkinson disease protein 7, protein DJ-1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Park7/DJ-1 (Q99497-1; Met1-Asp189).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cancer, DNA replication Transcription Translation and Splicing, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 11315
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.