missing translation for 'onlineSavingsMsg'
Learn More

Paricalcitol-d6, MedChemExpress

Product Code. 16481969
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16481969

Brand: Medchem Express HY76585

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Paricalcitol-d6 is the deuterium labeled Paricalcitol. Paricalcitol is a drug used for the prevention and treatment of secondary hyperparathyroidism (excessive secretion of parathyroid hormone) associated with chronic renal failure.

Paricalcitol-d6 is the deuterium labeled Paricalcitol. Paricalcitol is a drug used for the prevention and treatment of secondary hyperparathyroidism (excessive secretion of parathyroid hormone) associated with chronic renal failure.

Specifications

Chemical Name or Material Paricalcitol-d6
CAS 2070009-67-3
Purity Grade Notes Research
Physical Form Solid
Molecular Formula C27H38D6O3
Color White
Solubility Information DMSO : 100 mg/mL (236.59 mM; Need ultrasonic)
SMILES C[C@]([C@@H]([C@@H](/C=C/[C@@H](C(C([2H])([2H])[2H])(C([2H])([2H])[2H])O)C)C)CC1)(CCC/2)[C@@]1(C2=C\C=C3C[C@H](C[C@@H](C/3)O)O)[H]
Industry Type Metabolism-sugar/lipid metabolism
Molecular Weight (g/mol) 422.67
Quantity 1 mg
Formula Weight 422.67
Percent Purity 99.64%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.