missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARG Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27808-1-AP-150UL
This item is not returnable.
View return policy
Description
Poly(ADP-ribose) glycohydrolase (PARG) is the major enzyme responsible for the catabolism of poly(ADP-ribose), a reversible covalent-modifier of chromosomal proteins. The protein is found in many tissues and may be subject to proteolysis generating smaller, active products.Specifications
| PARG | |
| Polyclonal | |
| Unconjugated | |
| PARG | |
| PARG, PARG99 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 8505 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.47 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q86W56 | |
| PARG | |
| PARG Fusion Protein Ag27137 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction