missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARG Antibody (OTI6F4), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73245PE
This item is not returnable.
View return policy
Description
PARG Monoclonal antibody specifically detects PARG in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| PARG | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 8505 | |
| Store at 4C in the dark. | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| OTI6F4 | |
| Western Blot, Immunohistochemistry | |
| EC 3.2.1.143, FLJ54459, FLJ60257, FLJ60456, PARG99, poly (ADP-ribose) glycohydrolase, poly(ADP-ribose) glycohydrolase, poly(ADP-ribose) glycohydrolase 60 kDa isoform | |
| Human recombinant protein fragment corresponding to amino acids 1-367 of human PARG(NP_003622) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction