missing translation for 'onlineSavingsMsg'
Learn More

PARD6B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications

£213.00 - £428.00

Specifications

Antigen PARD6B
Dilution Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50, Chromatin Immunoprecipitation (ChIP)
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ChIP Assay
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18425491
View Documents
Novus Biologicals
NBP1-87337-25ul
25 μL
£213.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18040794
View Documents
Novus Biologicals
NBP1-87337
0.1 mL
£428.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PARD6B Polyclonal specifically detects PARD6B in Human, Mouse samples. It is validated for Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

PARD6B
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ChIP Assay
Unconjugated
RUO
Human
par-6 (partitioning defective 6, C.elegans) homolog beta, PAR-6 beta, par-6 partitioning defective 6 homolog beta (C. elegans), PAR6B, PAR-6B, partitioning defective 6 homolog beta
PARD6B
IgG
Affinity Purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50, Chromatin Immunoprecipitation (ChIP)
Polyclonal
Rabbit
Cell Cycle and Replication
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
84612
This antibody was developed against Recombinant Protein corresponding to amino acids:NYHKAVSTANPLLRIFIQKKEEADYSAFGTDTLIKKKNVLTNVLRPDNHRKKPHIVISMPQDFRPVSSIIDVDILPETHRRV
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.